Product Details

SNP ID
rs150170942
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11394285 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGGGTCCAATGGGCTACAGTTG[A/G]GTGGTGGTCCTGGGATACACAGCAC
Phenotype
MIM: 133171 MIM: 616743
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EPOR PubMed Links

Gene Details

Gene
EPOR
Gene Name
erythropoietin receptor
There are no transcripts associated with this gene.

Gene
RGL3
Gene Name
ral guanine nucleotide dissociation stimulator like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001035223.3 2332 UTR 3 NP_001030300.3
NM_001161616.2 2332 UTR 3 NP_001155088.2

View Full Product Details