Product Details

SNP ID
rs150234605
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40742346 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGACCCCTGAGACCTAGCTTCA[A/G]TCTTGTCGCCATCACACAGTGGGAG
Phenotype
MIM: 606476 MIM: 182285
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C19orf54 PubMed Links

Gene Details

Gene
C19orf54
Gene Name
chromosome 19 open reading frame 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198476.3 1719 UTR 3 NP_940878.3
XM_005258776.4 1719 UTR 3 XP_005258833.1
XM_005258778.4 1719 UTR 3 XP_005258835.1
XM_006723152.2 1719 Intron XP_006723215.1
XM_006723153.2 1719 Intron XP_006723216.1
XM_011526774.2 1719 Intron XP_011525076.1
XM_011526775.1 1719 Intron XP_011525077.1
XM_011526776.2 1719 Intron XP_011525078.1
XM_011526777.2 1719 Intron XP_011525079.1
XM_011526778.2 1719 Intron XP_011525080.1
XM_011526779.2 1719 Intron XP_011525081.1
XM_011526782.2 1719 Intron XP_011525084.1
XM_011526783.2 1719 Intron XP_011525085.1
XM_011526784.2 1719 Intron XP_011525086.1
XM_011526785.2 1719 Intron XP_011525087.1
XM_011526786.2 1719 Intron XP_011525088.1
XM_017026620.1 1719 Intron XP_016882109.1
XM_017026621.1 1719 Intron XP_016882110.1
XM_017026622.1 1719 Intron XP_016882111.1
XM_017026623.1 1719 Intron XP_016882112.1
XM_017026624.1 1719 Intron XP_016882113.1
XM_017026625.1 1719 Intron XP_016882114.1
XM_017026626.1 1719 UTR 3 XP_016882115.1
XM_017026627.1 1719 UTR 3 XP_016882116.1
Gene
ITPKC
Gene Name
inositol-trisphosphate 3-kinase C
There are no transcripts associated with this gene.

Gene
SNRPA
Gene Name
small nuclear ribonucleoprotein polypeptide A
There are no transcripts associated with this gene.

View Full Product Details