Product Details

SNP ID
rs150535617
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11377480 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAACAGGGGATCCATCTAAGTAC[C/T]CTAAGAGAAGGTAGAGCTACAGACT
Phenotype
MIM: 133171 MIM: 614536
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EPOR PubMed Links

Gene Details

Gene
EPOR
Gene Name
erythropoietin receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000121.3 2166 UTR 3 NP_000112.1
Gene
SWSAP1
Gene Name
SWIM-type zinc finger 7 associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175871.3 2166 Intron NP_787067.2

View Full Product Details