Product Details

SNP ID
rs150830749
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43203995 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCTGCGTGACATTCTGGATCAG[C/G]AGGGATGCATTGGAATATACTCTTT
Phenotype
MIM: 176393
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC284344 PubMed Links

Gene Details

Gene
LOC284344
Gene Name
uncharacterized LOC284344
There are no transcripts associated with this gene.

Gene
PSG4
Gene Name
pregnancy specific beta-1-glycoprotein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276495.1 559 Silent Mutation CTC,CTG L107L NP_001263424.1
NM_001316339.1 559 Intron NP_001303268.1
NM_002780.4 559 Silent Mutation CTC,CTG L107L NP_002771.2
NM_213633.2 559 Silent Mutation CTC,CTG L107L NP_998798.1
XM_017026995.1 559 Intron XP_016882484.1
XM_017026996.1 559 Silent Mutation CTC,CTG L123L XP_016882485.1
XM_017026997.1 559 Silent Mutation CTC,CTG L107L XP_016882486.1
XM_017026998.1 559 Intron XP_016882487.1
XM_017026999.1 559 Intron XP_016882488.1
XM_017027000.1 559 Silent Mutation CTC,CTG L123L XP_016882489.1
XM_017027001.1 559 Intron XP_016882490.1
XM_017027002.1 559 Intron XP_016882491.1

View Full Product Details