Product Details

SNP ID
rs150942017
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35633088 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACACCCTGACAGATTCCGCTCTC[G/T]CCTCTGCAGCAGCCGGCCCCCGCCC
Phenotype
MIM: 609358 MIM: 613432 MIM: 613232
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ETV2 PubMed Links

Gene Details

Gene
ETV2
Gene Name
ETS variant 2
There are no transcripts associated with this gene.

Gene
HAUS5
Gene Name
HAUS augmin like complex subunit 5
There are no transcripts associated with this gene.

Gene
RBM42
Gene Name
RNA binding motif protein 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319113.1 658 Intron NP_001306042.1
NM_024321.4 658 Missense Mutation GCC,TCC A174S NP_077297.2
XM_005259257.2 658 Intron XP_005259314.1

View Full Product Details