Product Details

SNP ID
rs112195274
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:37997873 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCACTCCACCCTGTGTCAGCGTCT[G/T]TGGGGGCAGCATCCATTAGTAGGTA
Phenotype
MIM: 602790 MIM: 605596
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FHL3 PubMed Links

Gene Details

Gene
FHL3
Gene Name
four and a half LIM domains 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243878.1 Intron NP_001230807.1
NM_004468.4 Intron NP_004459.2
XM_017000675.1 Intron XP_016856164.1
Gene
SF3A3
Gene Name
splicing factor 3a subunit 3
There are no transcripts associated with this gene.

View Full Product Details