Product Details
- SNP ID
-
rs112262020
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
11
- Location
-
Chr.1:155054396 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTTCAGGGTCCTCCCATTATTTCG[A/C]GAATCCAAGATCTCCACCTGCCAGG
- Phenotype
-
MIM: 605548
MIM: 601380
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ADAM15
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11548311] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADAM15
- Gene Name
- ADAM metallopeptidase domain 15
- Gene
- DCST1
- Gene Name
- DC-STAMP domain containing 1
There are no transcripts associated with this gene.
- Gene
- EFNA4
- Gene Name
- ephrin A4
There are no transcripts associated with this gene.
- Gene
- LOC100505666
- Gene Name
- uncharacterized LOC100505666
There are no transcripts associated with this gene.
View Full Product Details