Product Details

SNP ID
rs112651655
Assay Type
Functionally Tested
NCBI dbSNP Submissions
2
Location
Chr.1:37997562 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACATACTTGCCTCCACCGAGTCCT[A/G]GAGGGAGGCTGGAAGTTAGCTATGC
Phenotype
MIM: 602790 MIM: 605596
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
FHL3 PubMed Links

Gene Details

Gene
FHL3
Gene Name
four and a half LIM domains 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243878.1 Intron NP_001230807.1
NM_004468.4 Intron NP_004459.2
XM_017000675.1 Intron XP_016856164.1
Gene
SF3A3
Gene Name
splicing factor 3a subunit 3
There are no transcripts associated with this gene.

View Full Product Details