Product Details

SNP ID
rs113010659
Assay Type
Functionally Tested
NCBI dbSNP Submissions
2
Location
Chr.1:114716917 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAATAATGAAAGCGCTAGGTGCT[A/T]GTGTCTCAAAAATCAGTAAAACTTT
Phenotype
MIM: 191510 MIM: 164790
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
CSDE1 PubMed Links

Gene Details

Gene
CSDE1
Gene Name
cold shock domain containing E1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007553.2 3775 UTR 3 NP_001007554.1
NM_001130523.2 3775 UTR 3 NP_001123995.1
NM_001242891.1 3775 UTR 3 NP_001229820.1
NM_001242892.1 3775 UTR 3 NP_001229821.1
NM_001242893.1 3775 UTR 3 NP_001229822.1
NM_007158.5 3775 UTR 3 NP_009089.4
Gene
NRAS
Gene Name
neuroblastoma RAS viral oncogene homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002524.4 3775 Intron NP_002515.1

View Full Product Details