Product Details

SNP ID
rs113542183
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:159171798 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCAGCCGCCTCGCTCCTGCTCCT[G/T]CTCCTGCTGTTCGCCTGCTGCTGGG
Phenotype
MIM: 609743
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CADM3 PubMed Links

Gene Details

Gene
CADM3
Gene Name
cell adhesion molecule 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127173.1 212 Silent Mutation CTG,CTT L11L NP_001120645.1
NM_021189.3 212 Silent Mutation CTG,CTT L11L NP_067012.1

View Full Product Details