Product Details

SNP ID
rs113562433
Assay Type
Functionally Tested
NCBI dbSNP Submissions
16
Location
Chr.1:210681438 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAGCCTGATGTGAGAAGCTACAT[A/G]TGAGGTTAGGGCTCATGGAGCAGGA
Phenotype
MIM: 605743 MIM: 603305
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
HHAT PubMed Links

Gene Details

Gene
HHAT
Gene Name
hedgehog acyltransferase
There are no transcripts associated with this gene.

Gene
KCNH1
Gene Name
potassium voltage-gated channel subfamily H member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002238.3 4983 UTR 3 NP_002229.1
NM_172362.2 4983 UTR 3 NP_758872.1
XM_017001246.1 4983 UTR 3 XP_016856735.1

View Full Product Details