Product Details

SNP ID
rs114198341
Assay Type
Functionally tested
NCBI dbSNP Submissions
26
Location
Chr.1:6244394 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAAAAAGCGCCGGGCACGCATCA[A/G]TGTGTCACTGGAGCAGCTCAAGTCG
Phenotype
MIM: 614269 MIM: 609971 MIM: 605851
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR153 PubMed Links

Gene Details

Gene
GPR153
Gene Name
G protein-coupled receptor 153
There are no transcripts associated with this gene.

Gene
HES3
Gene Name
hes family bHLH transcription factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024598.3 94 Missense Mutation AAT,AGT N10S NP_001019769.1
Gene
ICMT
Gene Name
isoprenylcysteine carboxyl methyltransferase
There are no transcripts associated with this gene.

Gene
LINC00337
Gene Name
long intergenic non-protein coding RNA 337
There are no transcripts associated with this gene.

View Full Product Details