Product Details

SNP ID
rs114880122
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:248521592 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTGACCTCATTACTTTCTATCC[C/T]CAAATATTAATCACTTAGTATTTGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR2G6 PubMed Links

Gene Details

Gene
OR2G6
Gene Name
olfactory receptor family 2 subfamily G member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013355.1 Intron NP_001013373.1
Gene
OR2T10
Gene Name
olfactory receptor family 2 subfamily T member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004693.1 Intron NP_001004693.1
XM_017000273.1 Intron XP_016855762.1

View Full Product Details