Product Details

SNP ID
rs115228148
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:20296374 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGATTGCACGGTGAAACTGACAT[C/T]ATGTGAAAGTTGTGGGAGTTAGTGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VWA5B1 PubMed Links

Gene Details

Gene
VWA5B1
Gene Name
von Willebrand factor A domain containing 5B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039500.2 Intron NP_001034589.2
XM_006710360.3 Intron XP_006710423.2
XM_011540683.2 Intron XP_011538985.1
XM_011540684.2 Intron XP_011538986.1
XM_011540685.2 Intron XP_011538987.1
XM_011540688.2 Intron XP_011538990.1
XM_011540690.2 Intron XP_011538992.1
XM_011540691.2 Intron XP_011538993.1
XM_011540692.2 Intron XP_011538994.1
XM_011540693.1 Intron XP_011538995.1
XM_011540695.1 Intron XP_011538997.1
XM_011540697.2 Intron XP_011538999.1
XM_011540698.1 Intron XP_011539000.1
XM_017000305.1 Intron XP_016855794.1
XM_017000306.1 Intron XP_016855795.1

View Full Product Details