Product Details
- SNP ID
-
rs115249920
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
12
- Location
-
Chr.1:175069878 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTCCTCCTTGCTAGAAGACTGGAA[A/C]TTTAAAGTTACTTCAATATGTGGAA
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TNN
PubMed Links
Gene Details
- Gene
- TNN
- Gene Name
- tenascin N
View Full Product Details