Product Details

SNP ID
rs115254209
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:32682861 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATGTACAACTTGACACGGACTTT[C/G]TTTTATCCTTAGTTTCTTTCACGGA
Phenotype
MIM: 602923 MIM: 611750
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RBBP4 PubMed Links

Gene Details

Gene
RBBP4
Gene Name
RB binding protein 4, chromatin remodeling factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135255.1 4493 UTR 3 NP_001128727.1
NM_001135256.1 4493 UTR 3 NP_001128728.1
NM_005610.2 4493 UTR 3 NP_005601.1
Gene
SYNC
Gene Name
syncoilin, intermediate filament protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161708.1 4493 Intron NP_001155180.1
NM_030786.2 4493 Intron NP_110413.2

View Full Product Details