Product Details

SNP ID
rs115279484
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:152349651 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTGGGTTAGTTCTTATCTTGGG[A/C]TAGTTTATGTTGGCTTCTTATTCAA
Phenotype
MIM: 616284
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FLG-AS1 PubMed Links

Gene Details

Gene
FLG-AS1
Gene Name
FLG antisense RNA 1
There are no transcripts associated with this gene.

Gene
FLG2
Gene Name
filaggrin family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014342.2 7774 UTR 3 NP_001014364.1
XM_011509531.2 7774 UTR 3 XP_011507833.1

View Full Product Details