Product Details

SNP ID
rs115385043
Assay Type
Functionally Tested
NCBI dbSNP Submissions
8
Location
Chr.1:35181903 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGTACATTGCTGTTTTAGTGAT[A/G]AAAGTCAAATGTCATTTGAAACTAA
Phenotype
MIM: 605199
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
SFPQ PubMed Links

Gene Details

Gene
SFPQ
Gene Name
splicing factor proline and glutamine rich
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005066.2 4789 Intron NP_005057.1
XM_005271112.4 4789 UTR 3 XP_005271169.1
XM_005271113.4 4789 UTR 3 XP_005271170.1
XM_005271115.4 4789 Intron XP_005271172.1
XM_011541950.2 4789 UTR 3 XP_011540252.1
XM_017002053.1 4789 UTR 3 XP_016857542.1
XM_017002054.1 4789 UTR 3 XP_016857543.1

View Full Product Details