Product Details
- SNP ID
-
rs115388123
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
18
- Location
-
Chr.1:11921245 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGGCACAGTAGGCAGCCTCACCCC[A/G]CGACAGCAATGCTGATGCTAAGATG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KIAA2013
PubMed Links
Gene Details
- Gene
- KIAA2013
- Gene Name
- KIAA2013
View Full Product Details