Product Details

SNP ID
rs116143898
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:15113310 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGGCGGTGGCAAAAATTCTTATT[C/G]TTTCTATGTAAAAGCACAAATGTAC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KAZN PubMed Links

Gene Details

Gene
KAZN
Gene Name
kazrin, periplakin interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017999.2 3276 Intron NP_001017999.1
NM_001018000.3 3276 Intron NP_001018000.1
NM_001018001.2 3276 Intron NP_001018001.1
NM_015209.2 3276 Intron NP_056024.1
NM_201628.2 3276 Intron NP_963922.2
XM_005245795.4 3276 Intron XP_005245852.1
XM_011541074.2 3276 UTR 3 XP_011539376.1
XM_011541075.2 3276 UTR 3 XP_011539377.1
XM_011541076.2 3276 UTR 3 XP_011539378.1
XM_011541077.2 3276 UTR 3 XP_011539379.1
XM_011541080.2 3276 Intron XP_011539382.1
XM_011541081.2 3276 UTR 3 XP_011539383.1
XM_017000768.1 3276 Intron XP_016856257.1
XM_017000769.1 3276 Intron XP_016856258.1
XM_017000770.1 3276 Intron XP_016856259.1
XM_017000771.1 3276 Intron XP_016856260.1
XM_017000772.1 3276 Intron XP_016856261.1
Gene
TMEM51-AS1
Gene Name
TMEM51 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details