Product Details

SNP ID
rs137942881
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:233905622 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAGTACTTGGTGGTGGACGAGGC[A/G]ATTAAGGAGGATCTGAAATGGTCGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC35F3 PubMed Links

Gene Details

Gene
SLC35F3
Gene Name
solute carrier family 35 member F3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300845.1 513 Intron NP_001287774.1
NM_173508.3 513 Silent Mutation GCA,GCG A49A NP_775779.1

View Full Product Details