Product Details

SNP ID
rs137955605
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:150284469 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGTGTGGCAATGGACAGGATCCA[A/G]CGTATTGTAGGTGTTTTGCAGAAGC
Phenotype
MIM: 607629 MIM: 615782 MIM: 611984
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APH1A PubMed Links

Gene Details

Gene
APH1A
Gene Name
aph-1 homolog A, gamma-secretase subunit
There are no transcripts associated with this gene.

Gene
C1orf54
Gene Name
chromosome 1 open reading frame 54
There are no transcripts associated with this gene.

Gene
CIART
Gene Name
circadian associated repressor of transcription
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300838.1 487 Silent Mutation CAA,CAG Q162Q NP_001287767.1
NM_001300839.1 487 Silent Mutation CAA,CAG Q74Q NP_001287768.1
NM_001300840.1 487 Silent Mutation CAA,CAG Q74Q NP_001287769.1
NM_001300841.1 487 Silent Mutation CAA,CAG Q74Q NP_001287770.1
NM_144697.3 487 Silent Mutation CAA,CAG Q162Q NP_653298.1
XM_017000378.1 487 Silent Mutation CAA,CAG Q5Q XP_016855867.1
Gene
MRPS21
Gene Name
mitochondrial ribosomal protein S21
There are no transcripts associated with this gene.

View Full Product Details