Product Details

SNP ID
rs138170081
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:156646971 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTTTCCTGTCCCGGGGCCGGGAG[G/T]CAGAGGTGCTGGTGGCGCGGGGAGT
Phenotype
MIM: 600347
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BCAN PubMed Links

Gene Details

Gene
BCAN
Gene Name
brevican
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021948.4 598 Missense Mutation GCA,TCA A88S NP_068767.3
NM_198427.1 598 Missense Mutation GCA,TCA A88S NP_940819.1
XM_011509866.1 598 Missense Mutation GCA,TCA A88S XP_011508168.1
XM_017002047.1 598 Missense Mutation GCA,TCA A133S XP_016857536.1

View Full Product Details