Product Details

SNP ID
rs138246571
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:247841032 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTCTTTGTTCCTCAAGCTGTAGA[C/T]AACTGGGTTCAGCAGTGGTGTGACC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR11L1 PubMed Links

Gene Details

Gene
OR11L1
Gene Name
olfactory receptor family 11 subfamily L member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001959.1 865 Missense Mutation NP_001001959.1

View Full Product Details