Product Details
- SNP ID
-
rs138326583
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:23024173 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAGATTTTTTTCTTTGTAATTCTC[A/C]TGTGTTTATTTTAACTCATCTTACT
- Phenotype
-
MIM: 609132
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C1orf234
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs138534510] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C1orf234
- Gene Name
- chromosome 1 open reading frame 234
There are no transcripts associated with this gene.
- Gene
- KDM1A
- Gene Name
- lysine demethylase 1A
View Full Product Details