Product Details

SNP ID
rs138443680
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:161750021 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCAAGGCGGGGCCTGGGGTCGAG[A/G]ATCTATGGCGCCTCTTCGTGCCAGC
Phenotype
MIM: 604835
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DUSP12 PubMed Links

Gene Details

Gene
DUSP12
Gene Name
dual specificity phosphatase 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007240.2 254 Missense Mutation AAT,GAT N74D NP_009171.1
XM_005244862.2 254 UTR 5 XP_005244919.1

View Full Product Details