Product Details
- SNP ID
-
rs138726845
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
8
- Location
-
Chr.1:237004261 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGGCACACTTGGCACAGCCCAGGG[A/G]GCAACAGGAGCAGCAGCTCTTCTTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MT1HL1
PubMed Links
Gene Details
- Gene
- MT1HL1
- Gene Name
- metallothionein 1H-like 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001276687.1 |
158 |
Missense Mutation |
CCC,TCC |
P38S |
NP_001263616.1 |
View Full Product Details