Product Details
- SNP ID
-
rs138805281
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
16
- Location
-
Chr.1:88857300 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCTCCTCCCAGGAACCAAGTCCAG[A/T]TCCACAGCTTTACGGGCTATATGTG
- Phenotype
-
MIM: 189963
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GTF2B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2794318] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GTF2B
- Gene Name
- general transcription factor IIB
View Full Product Details