Product Details

SNP ID
rs138818377
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:151176711 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGGGAGGGGCTCAGGAGGTGGC[A/G]GGCCGGGGCCCAGGGCTGAGGCAGT
Phenotype
MIM: 608095 MIM: 605834 MIM: 600607
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SCNM1 PubMed Links

Gene Details

Gene
SCNM1
Gene Name
sodium channel modifier 1
There are no transcripts associated with this gene.

Gene
TMOD4
Gene Name
tropomodulin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013353.2 1120 Intron NP_037485.2
XM_011509449.1 1120 Intron XP_011507751.1
XM_017001089.1 1120 Intron XP_016856578.1
XM_017001090.1 1120 Intron XP_016856579.1
Gene
TNFAIP8L2-SCNM1
Gene Name
TNFAIP8L2-SCNM1 readthrough
There are no transcripts associated with this gene.

Gene
VPS72
Gene Name
vacuolar protein sorting 72 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271087.1 1120 Missense Mutation CCG,CTG P354L NP_001258016.1
NM_001271088.1 1120 UTR 3 NP_001258017.1
NM_005997.2 1120 Missense Mutation CCG,CTG P343L NP_005988.1
XM_017002205.1 1120 Missense Mutation CCG,CTG P222L XP_016857694.1

View Full Product Details