Product Details

SNP ID
rs138830904
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:15112874 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCTGGAAAAGGAGTTCTCTGGG[A/G]CGAGGGGGATCCTGCATTCAGCTAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KAZN PubMed Links

Gene Details

Gene
KAZN
Gene Name
kazrin, periplakin interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017999.2 2840 Intron NP_001017999.1
NM_001018000.3 2840 Intron NP_001018000.1
NM_001018001.2 2840 Intron NP_001018001.1
NM_015209.2 2840 Intron NP_056024.1
NM_201628.2 2840 Intron NP_963922.2
XM_005245795.4 2840 Intron XP_005245852.1
XM_011541074.2 2840 Silent Mutation GGA,GGG G930G XP_011539376.1
XM_011541075.2 2840 Silent Mutation GGA,GGG G826G XP_011539377.1
XM_011541076.2 2840 Silent Mutation GGA,GGG G738G XP_011539378.1
XM_011541077.2 2840 Silent Mutation GGA,GGG G738G XP_011539379.1
XM_011541080.2 2840 Intron XP_011539382.1
XM_011541081.2 2840 Silent Mutation GGA,GGG G473G XP_011539383.1
XM_017000768.1 2840 Intron XP_016856257.1
XM_017000769.1 2840 Intron XP_016856258.1
XM_017000770.1 2840 Intron XP_016856259.1
XM_017000771.1 2840 Intron XP_016856260.1
XM_017000772.1 2840 Intron XP_016856261.1
Gene
TMEM51-AS1
Gene Name
TMEM51 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details