Product Details

SNP ID
rs139006399
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:206410286 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTGCCTCTTATTTGCCCTCAGT[A/G]GTGACATGTTTCCTGATCTTCTTGG
Phenotype
MIM: 606524
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SRGAP2 PubMed Links

Gene Details

Gene
SRGAP2
Gene Name
SLIT-ROBO Rho GTPase activating protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170637.3 Intron NP_001164108.1
NM_001300952.1 Intron NP_001287881.1
NM_015326.4 Intron NP_056141.2
XM_005277510.2 Intron XP_005277567.1
XM_005277511.2 Intron XP_005277568.1
XM_005277514.1 Intron XP_005277571.1
XM_005277515.1 Intron XP_005277572.1
XM_011509354.1 Intron XP_011507656.1
XM_011509355.1 Intron XP_011507657.1
XM_011509356.2 Intron XP_011507658.1
XM_011509359.1 Intron XP_011507661.1
XM_017000838.1 Intron XP_016856327.1
XM_017000839.1 Intron XP_016856328.1
XM_017000840.1 Intron XP_016856329.1
XM_017000841.1 Intron XP_016856330.1

View Full Product Details