Product Details

SNP ID
rs139152573
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:1787370 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGAAGCTATCCCAGGACCCTGT[C/T]GCCACAGCCATGCCATCGTCAGTCA
Phenotype
MIM: 139380 MIM: 611616
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GNB1 PubMed Links

Gene Details

Gene
GNB1
Gene Name
G protein subunit beta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282538.1 1281 Silent Mutation GCA,GCG A228A NP_001269467.1
NM_001282539.1 1281 Silent Mutation GCA,GCG A328A NP_001269468.1
NM_002074.4 1281 Silent Mutation GCA,GCG A328A NP_002065.1
XM_017001059.1 1281 Silent Mutation GCA,GCG A328A XP_016856548.1
XM_017001060.1 1281 Silent Mutation GCA,GCG A328A XP_016856549.1
XM_017001061.1 1281 Silent Mutation GCA,GCG A328A XP_016856550.1
Gene
NADK
Gene Name
NAD kinase
There are no transcripts associated with this gene.

View Full Product Details