Product Details

SNP ID
rs139334642
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:46310170 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTACATCAGGATCCCCTAACAACA[C/G]TGAGAGAGCAATGCGAGCAGTTGGA
Phenotype
MIM: 613844
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LRRC41 PubMed Links

Gene Details

Gene
LRRC41
Gene Name
leucine rich repeat containing 41
There are no transcripts associated with this gene.

Gene
UQCRH
Gene Name
ubiquinol-cytochrome c reductase hinge protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297565.1 251 Missense Mutation CTG,GTG L27V NP_001284494.1
NM_001297566.1 251 Missense Mutation CTG,GTG L24V NP_001284495.1
NM_006004.3 251 Missense Mutation CTG,GTG L33V NP_005995.2

View Full Product Details