Product Details
- SNP ID
-
rs139504725
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:156937269 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCACCTTGAGCCTGTTGAGCTTGA[A/G]AGTGAGCTGCTCAATGGTATGGAAG
- Phenotype
-
MIM: 605708
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ARHGEF11
PubMed Links
Gene Details
- Gene
- ARHGEF11
- Gene Name
- Rho guanine nucleotide exchange factor 11
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014784.3 |
3928 |
Missense Mutation |
CTC,TTC |
L1434F |
NP_055599.1 |
NM_198236.2 |
3928 |
Missense Mutation |
CTC,TTC |
L1474F |
NP_937879.1 |
XM_005245629.4 |
3928 |
Missense Mutation |
CTC,TTC |
L1475F |
XP_005245686.1 |
XM_005245633.4 |
3928 |
Missense Mutation |
CTC,TTC |
L1053F |
XP_005245690.1 |
XM_006711659.2 |
3928 |
Missense Mutation |
CTC,TTC |
L1491F |
XP_006711722.1 |
XM_006711660.3 |
3928 |
Missense Mutation |
CTC,TTC |
L1490F |
XP_006711723.1 |
XM_006711661.2 |
3928 |
Missense Mutation |
CTC,TTC |
L1481F |
XP_006711724.1 |
XM_006711662.2 |
3928 |
Missense Mutation |
CTC,TTC |
L1480F |
XP_006711725.1 |
XM_006711663.3 |
3928 |
Missense Mutation |
CTC,TTC |
L1471F |
XP_006711726.1 |
XM_006711665.3 |
3928 |
Missense Mutation |
CTC,TTC |
L1450F |
XP_006711728.1 |
XM_011510185.2 |
3928 |
Missense Mutation |
CTC,TTC |
L1486F |
XP_011508487.1 |
XM_011510186.2 |
3928 |
Missense Mutation |
CTC,TTC |
L1451F |
XP_011508488.1 |
XM_011510187.2 |
3928 |
Missense Mutation |
CTC,TTC |
L1397F |
XP_011508489.1 |
XM_011510189.2 |
3928 |
Missense Mutation |
CTC,TTC |
L857F |
XP_011508491.1 |
XM_017002920.1 |
3928 |
Missense Mutation |
CTC,TTC |
L1480F |
XP_016858409.1 |
XM_017002921.1 |
3928 |
Missense Mutation |
CTC,TTC |
L1464F |
XP_016858410.1 |
XM_017002922.1 |
3928 |
Missense Mutation |
CTC,TTC |
L1440F |
XP_016858411.1 |
XM_017002923.1 |
3928 |
Missense Mutation |
CTC,TTC |
L1435F |
XP_016858412.1 |
XM_017002924.1 |
3928 |
Missense Mutation |
CTC,TTC |
L1435F |
XP_016858413.1 |
- Gene
- LRRC71
- Gene Name
- leucine rich repeat containing 71
- Gene
- MIR765
- Gene Name
- microRNA 765
There are no transcripts associated with this gene.
View Full Product Details