Product Details

SNP ID
rs139672928
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:9932849 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGTTAGTATCTCCACTTTCTGT[G/T]GAGTGTACAGGTCTCTTTCCAGCTT
Phenotype
MIM: 610458
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LZIC PubMed Links

Gene Details

Gene
LZIC
Gene Name
leucine zipper and CTNNBIP1 domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316973.1 732 Missense Mutation CAA,CCA Q129P NP_001303902.1
NM_001316974.1 732 Missense Mutation CAA,CCA Q150P NP_001303903.1
NM_001316975.1 732 Missense Mutation CAA,CCA Q129P NP_001303904.1
NM_001316976.1 732 Intron NP_001303905.1
NM_032368.4 732 Missense Mutation CAA,CCA Q129P NP_115744.2
XM_005263506.2 732 Missense Mutation CAA,CCA Q129P XP_005263563.1
XM_005263509.2 732 Intron XP_005263566.1
XM_017002546.1 732 Missense Mutation CAA,CCA Q129P XP_016858035.1
XM_017002547.1 732 Missense Mutation CAA,CCA Q129P XP_016858036.1

View Full Product Details