Product Details

SNP ID
rs139894605
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:161983781 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCTGGGTCGTATAGGAATACTCA[G/T]TCTCGAACAGCAGCCTGGGGACGAT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OLFML2B PubMed Links

Gene Details

Gene
OLFML2B
Gene Name
olfactomedin like 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297713.1 3188 Missense Mutation NP_001284642.1
NM_015441.2 3188 Missense Mutation NP_056256.1
XM_005245075.2 3188 Missense Mutation XP_005245132.1
XM_011509398.2 3188 Missense Mutation XP_011507700.1
XM_017000967.1 3188 Intron XP_016856456.1

View Full Product Details