Product Details

SNP ID
rs140029453
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:40988627 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCGTGGGGGACATAGAAAGCATG[C/T]CCTTTATTGAGGCCTTCCGTCAGTT
Phenotype
MIM: 123860
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTPS1 PubMed Links

Gene Details

Gene
CTPS1
Gene Name
CTP synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301237.1 1016 Missense Mutation CCC,TCC P2S NP_001288166.1
NM_001905.3 1016 Missense Mutation CCC,TCC P158S NP_001896.2

View Full Product Details