Product Details

SNP ID
rs140067092
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:158398889 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATAAAGCCAGCTCTTTCACATGG[G/T]TGTCAGTGCAGGCTAACTTGATAAC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR10T2 PubMed Links

Gene Details

Gene
OR10T2
Gene Name
olfactory receptor family 10 subfamily T member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004475.1 578 Missense Mutation AAC,ACC N193T NP_001004475.1

View Full Product Details