Product Details

SNP ID
rs140114227
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:229326857 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTTTCTTTTATTTTACTCGATG[A/G]TCTTTGGGGACTTTTGACCGCTTTC
Phenotype
MIM: 616762
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCSAP PubMed Links

Gene Details

Gene
CCSAP
Gene Name
centriole, cilia and spindle associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145257.4 595 Missense Mutation CCA,TCA P173S NP_660300.3

View Full Product Details