Product Details

SNP ID
rs140119973
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:161678144 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCTTCAGTGATGAGACTCTTAG[C/G]AAGGCAGGAATGGATAGGATAGGGG
Phenotype
MIM: 604590
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FCGR2B PubMed Links

Gene Details

Gene
FCGR2B
Gene Name
Fc fragment of IgG receptor IIb
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002273.2 1651 UTR 3 NP_001002273.1
NM_001002274.2 1651 UTR 3 NP_001002274.1
NM_001002275.2 1651 UTR 3 NP_001002275.1
NM_001190828.1 1651 UTR 3 NP_001177757.1
NM_004001.4 1651 UTR 3 NP_003992.3
XM_011509292.2 1651 Intron XP_011507594.1
XM_017000669.1 1651 Intron XP_016856158.1
XM_017000670.1 1651 Intron XP_016856159.1
XM_017000671.1 1651 UTR 3 XP_016856160.1
XM_017000672.1 1651 UTR 3 XP_016856161.1
Gene
RPL31P11
Gene Name
ribosomal protein L31 pseudogene 11
There are no transcripts associated with this gene.

View Full Product Details