Product Details

SNP ID
rs140142134
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:153630564 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACGGCGATGGAGACCCTCATCAAC[A/G]TGTTCCACGCCCACTCGGGCAAAGA
Phenotype
MIM: 614206 MIM: 176940 MIM: 601989
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHTOP PubMed Links

Gene Details

Gene
CHTOP
Gene Name
chromatin target of PRMT1
There are no transcripts associated with this gene.

Gene
S100A1
Gene Name
S100 calcium binding protein A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006271.1 156 Missense Mutation ATG,GTG M15V NP_006262.1
Gene
S100A13
Gene Name
S100 calcium binding protein A13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024210.1 156 Intron NP_001019381.1
NM_001024211.1 156 Intron NP_001019382.1
NM_001024212.1 156 Intron NP_001019383.1
NM_001024213.1 156 Intron NP_001019384.1
NM_005979.2 156 Intron NP_005970.1
XM_005245434.3 156 Intron XP_005245491.1
XM_011509862.2 156 Intron XP_011508164.1
XM_011509863.2 156 Intron XP_011508165.1
XM_011509864.1 156 UTR 5 XP_011508166.1
XM_017002034.1 156 Intron XP_016857523.1
XM_017002035.1 156 Intron XP_016857524.1
XM_017002036.1 156 Intron XP_016857525.1

View Full Product Details