Product Details

SNP ID
rs140375229
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:27382503 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGAGGGCTCAGCTCCATACCTG[G/T]CTCCTCTGGCCGGCACTGCTCCCAC
Phenotype
MIM: 604973
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CD164L2 PubMed Links

Gene Details

Gene
CD164L2
Gene Name
CD164 molecule like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005245868.3 329 Missense Mutation XP_005245925.2
XM_011541441.1 329 Missense Mutation XP_011539743.1
Gene
FCN3
Gene Name
ficolin 3
There are no transcripts associated with this gene.

View Full Product Details