Product Details

SNP ID
rs140454621
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:15112147 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGAGTTCAGATCTTGATTCTGCC[A/G]CTAACAGCTGTGTCGTCTTGGGCAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KAZN PubMed Links

Gene Details

Gene
KAZN
Gene Name
kazrin, periplakin interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017999.2 Intron NP_001017999.1
NM_001018000.3 Intron NP_001018000.1
NM_001018001.2 Intron NP_001018001.1
NM_015209.2 Intron NP_056024.1
NM_201628.2 Intron NP_963922.2
XM_005245795.4 Intron XP_005245852.1
XM_011541074.2 Intron XP_011539376.1
XM_011541075.2 Intron XP_011539377.1
XM_011541076.2 Intron XP_011539378.1
XM_011541077.2 Intron XP_011539379.1
XM_011541080.2 Intron XP_011539382.1
XM_011541081.2 Intron XP_011539383.1
XM_017000768.1 Intron XP_016856257.1
XM_017000769.1 Intron XP_016856258.1
XM_017000770.1 Intron XP_016856259.1
XM_017000771.1 Intron XP_016856260.1
XM_017000772.1 Intron XP_016856261.1
Gene
TMEM51-AS1
Gene Name
TMEM51 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details