Product Details

SNP ID
rs140628166
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:152350822 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTGAAAATGACTTGCTCTACTAG[A/G]TCTGGAACCTGTCTGTGTGGATTGT
Phenotype
MIM: 616284
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FLG-AS1 PubMed Links

Gene Details

Gene
FLG-AS1
Gene Name
FLG antisense RNA 1
There are no transcripts associated with this gene.

Gene
FLG2
Gene Name
filaggrin family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014342.2 6603 Missense Mutation CCT,TCT P2322S NP_001014364.1
XM_011509531.2 6603 Missense Mutation CCT,TCT P2170S XP_011507833.1

View Full Product Details