Product Details

SNP ID
rs140720320
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:152107590 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTTCTTCCCGTAATTTCCTTTCC[C/T]GTTCCTGGCGACGTTTCTGCTCCTC
Phenotype
MIM: 190370
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TCHH PubMed Links

Gene Details

Gene
TCHH
Gene Name
trichohyalin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007113.3 5722 Missense Mutation CAG,CGG Q1876R NP_009044.2

View Full Product Details