Product Details

SNP ID
rs140726188
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:196779193 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTGATTTTCCAGACATTAAACA[C/T]GGAGGTCTATTTCATGAGAATATGC
Phenotype
MIM: 605336
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CFHR3 PubMed Links

Gene Details

Gene
CFHR3
Gene Name
complement factor H related 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166624.1 177 Silent Mutation CAC,CAT H30H NP_001160096.1
NM_021023.5 177 Silent Mutation CAC,CAT H30H NP_066303.2

View Full Product Details