Product Details

SNP ID
rs140779257
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:235979106 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAGATCGAGAGCAACCACGGAAT[C/T]CCTACAAAGCACCAAAGGGCAGAAG
Phenotype
MIM: 131390
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NID1 PubMed Links

Gene Details

Gene
NID1
Gene Name
nidogen 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002508.2 3613 Missense Mutation AAT,GAT N1171D NP_002499.2
XM_011544195.2 3613 Missense Mutation AAT,GAT N1129D XP_011542497.1

View Full Product Details