Product Details

SNP ID
rs140866034
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:945084 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCGAGTTGCTGCTGTCCTCCTCGC[C/T]CTCCTCCTCGTCCTCTTCATCGTCT
Phenotype
MIM: 610770 MIM: 616765
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NOC2L PubMed Links

Gene Details

Gene
NOC2L
Gene Name
NOC2 like nucleolar associated transcriptional repressor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015658.3 Intron NP_056473.2
Gene
SAMD11
Gene Name
sterile alpha motif domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152486.2 Intron NP_689699.2

View Full Product Details