Product Details

SNP ID
rs141117966
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:26318024 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACGAAGATCCTACCAAAATGAAG[C/T]GCTTCCTCTTCCTCCTACTCACCAT
Phenotype
MIM: 114280 MIM: 609151
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AIM1L PubMed Links

Gene Details

Gene
AIM1L
Gene Name
absent in melanoma 1-like
There are no transcripts associated with this gene.

Gene
CD52
Gene Name
CD52 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001803.2 105 Missense Mutation CGC,TGC R3C NP_001794.2
Gene
UBXN11
Gene Name
UBX domain protein 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077262.1 105 Intron NP_001070730.1
NM_145345.2 105 Intron NP_663320.2
NM_183008.2 105 Intron NP_892120.2

View Full Product Details