Product Details

SNP ID
rs141468230
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:92246602 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATGAGTTCTTCCACTCGAGCTAG[A/G]ACACTTTCAATCAAATCAAATGTGA
Phenotype
MIM: 601749
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf146 PubMed Links

Gene Details

Gene
C1orf146
Gene Name
chromosome 1 open reading frame 146
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012425.1 1714 Intron NP_001012425.1
XM_011541447.2 1714 Intron XP_011539749.1
Gene
GLMN
Gene Name
glomulin, FKBP associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319683.1 1714 Silent Mutation NP_001306612.1
NM_053274.2 1714 Silent Mutation NP_444504.1
XM_005270401.3 1714 Silent Mutation XP_005270458.1
XM_006710309.2 1714 Silent Mutation XP_006710372.1
XM_011540546.2 1714 Silent Mutation XP_011538848.1
XM_017000137.1 1714 Silent Mutation XP_016855626.1
XM_017000138.1 1714 Silent Mutation XP_016855627.1
XM_017000139.1 1714 Silent Mutation XP_016855628.1
XM_017000140.1 1714 Silent Mutation XP_016855629.1
XM_017000141.1 1714 Silent Mutation XP_016855630.1
XM_017000142.1 1714 Silent Mutation XP_016855631.1
XM_017000143.1 1714 Silent Mutation XP_016855632.1
XM_017000144.1 1714 Silent Mutation XP_016855633.1

View Full Product Details